Learn More
Abeba™ Arrow 38200 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
Brand:  Abeba™ 38200/46
 View more versions of this product
Product Code. 15611148
  • £69.00 / 1 pair
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRA
  • Smooth leather white
  • Slip resistance PU/PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Microfiber heel patch
  • Lining microfiber, respiration absorbing
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRA
Microfiber
46
Polyurethane
White
Leather
Slip-resistant
38200
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Abeba™ Arrow 38200 Shoes > Size: 46