Learn More
Abeba™ Autoclavable Clogs 39600
Antistatic and autoclavable at 121°C. High slip resistance.
Brand: Abeba™ 39600/40
Description
- CE EN ISO 20347:2012 OB A, E SRC
- Autoclavable, sterilisable special material
- White
- Slip resistance outsole sole
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
- DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
CE EN ISO 20347:2012 OB A, E SRC | |
40 | |
39600 |
White | |
Slip-resistant |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Abeba™ Autoclavable Clogs 39600 > Size: 40
Spot an opportunity for improvement?Share a Content Correction