Learn More
Abeba™ Autoclavable Clogs 39600
Antistatic and autoclavable at 121°C. High slip resistance.
Brand:  Abeba™ 39600/38
 View more versions of this product
Product Code. 15621238
  • £56.00 / 1 pair
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • White
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRC
38
39600
White
Slip-resistant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Abeba™ Autoclavable Clogs 39600 > Size: 38