Learn More
Abeba™ Arrow 8210 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
Brand: Abeba™ 8210/41
Additional Details : Weight : 0.60000kg
Description
- CE EN ISO 20347:2012 OB A, E, FO SRA
- Smooth leather
- Black
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
- Microfiber heel patch
- Lining microfiber, respiration absorbing
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRA | |
Microfiber | |
41 | |
Polyurethane |
Black | |
Leather | |
Slip-resistant | |
8210 |