Learn More
Abeba™ Arrow 8210 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
Brand:  Abeba™ 8210/44
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 15693058
  • £61.53 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRA
  • Smooth leather
  • Black
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Microfiber heel patch
  • Lining microfiber, respiration absorbing
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRA
Microfiber
44
Polyurethane
Black
Leather
Slip-resistant
8210