missing translation for 'onlineSavingsMsg'
Learn More

Notrax Arrow Trax™ Non-Directional Chevron Patterned Entrance Mat

Notrax Entrance Mat - The needle punched fibers are durable and crush-resistant for extreme grime retention making it suitable for heavy traffic areas. Dense vinyl backing is moisture resistant and slip resistant to keep the mat in place.

Brand:  Notrax 118S0035CH

 View more versions of this product

Product Code. 17741938

  • £85.85 / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The non-directional chevron pattern of the Arrow Trax™ ensures a very aggressive scraping action making it one of our most popular carpet mats offering tremendous value
  • The needle punched fibers are durable and crush-resistant for extreme grime retention making it suitable for heavy traffic areas
  • Dense vinyl backing is moisture resistant and slip resistant to keep the mat in place
Specifications

Specifications

Charcoal
150 cm
Industrial and Commercial Matting
59 in.
Polyester
90 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.