Learn More
Arsenic(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
£43.40 - £638.00
Chemical Identifiers
CAS | 1327-53-3 |
---|---|
Molecular Formula | As2O3 |
Molecular Weight (g/mol) | 197.84 |
MDL Number | MFCD00003433 |
InChI Key | IKWTVSLWAPBBKU-UHFFFAOYSA-N |
IUPAC Name | diarsorosooxidane |
SMILES | O=[As]O[As]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363938
|
Thermo Scientific Alfa Aesar
040370.09 |
10 g |
£43.40
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343938
|
Thermo Scientific Alfa Aesar
040370.18 |
50 g |
£149.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353938
|
Thermo Scientific Alfa Aesar
040370.30 |
250 g |
£638.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1327-53-3 | |
197.84 | |
IKWTVSLWAPBBKU-UHFFFAOYSA-N | |
O=[As]O[As]=O |
As2O3 | |
MFCD00003433 | |
diarsorosooxidane |
Specifications
465°C (sublimation) | |
1327-53-3 | |
Powder | |
As2O3 | |
Soluble in dilute acids and alkalies.Insoluble in organic solvents.Slightly soluble in water. | |
O=[As]O[As]=O | |
197.84 | |
Odorless | |
Arsenic(III) oxide |
312.3°C | |
White | |
99.99% (Metals basis) | |
MFCD00003433 | |
IKWTVSLWAPBBKU-UHFFFAOYSA-N | |
diarsorosooxidane | |
197.84 | |
3.74 |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H300-H314-H335-H350-H371
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 215-481-4
missing translation for 'rtecsNumber' : CG3325000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only