missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals

£27.64 - £345.72

Chemical Identifiers

CAS 1327-53-3
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326188
View Documents
Thermo Scientific Alfa Aesar
011173.04
2 g
£27.64
2g
Please to purchase this item. Need a web account? Register with us today!
11346188
View Documents
Thermo Scientific Alfa Aesar
011173.09
10 g
£80.50
10g
Please to purchase this item. Need a web account? Register with us today!
11336188
View Documents
Thermo Scientific Alfa Aesar
011173.18
50 g
£345.72
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Arsenic trioxide is used as a precursor to forestry products, in colorless glass production, and in electronics. And it is also used as pigments, and preservatives for hides and wood, in the semiconductor industry to create light emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1327-53-3
197.84
IKWTVSLWAPBBKU-UHFFFAOYSA-N
O=[As]O[As]=O
As2O3
MFCD00003433
diarsorosooxidane

Specifications

465°C (sublimation)
1327-53-3
As2O3
UN1561
Soluble in hot water,dilute acids and alkalies. Insoluble in alcohol,chloroform,and ether,.
O=[As]O[As]=O
197.84
99.996%
Odorless
(metals basis excluding Sb), Sb 20ppm max.
312.3°C
Powder
MFCD00003433
14,804
IKWTVSLWAPBBKU-UHFFFAOYSA-N
diarsorosooxidane
197.84
Puratronic™
3.74 g/mL
Arsenic(III) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H350-H314-H318
Fatal if swallowed.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300-H314-H335-H350-H371

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC TRIOXIDE

missing translation for 'einecsNumber' : 215-481-4

missing translation for 'rtecsNumber' : CG3325000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only