missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts Aspergillus Double Gel Diffusion Assay
Microgen Bioproducts Aspergillus - A serological double gel diffusion assay intended for the detection of precipitating antibodies to Aspergillus fumigatus.
Brand: Novacyt - Microgen Bioproducts FSK1
This item is not returnable.
View return policy
Description
- Double diffusion assay for the detection of precipitating antibodies to Aspergillus fumigates in serum.
Specifications
8.5 mL | |
1 Kit | |
Double Gel Diffusion Assay | |
EC/REP label on kit box, Blank CE Carton label |
Liquid | |
Double gel diffusion (Ouchterlony) | |
Test Kit | |
5 to 10 mL of venous blood |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novacyt - Microgen Bioproducts Aspergillus Double Gel Diffusion Assay > Aspergillus Double Gel Diffusion Assay
Spot an opportunity for improvement?Share a Content Correction