Learn More
ATF4 Mouse anti-Human, Mouse, Clone: 3E4C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60035-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is a mouse monoclonal antibody raised against full length human ATF4 antigen. The antibody recognizes the 38 kDa ATF4 protein and its phosphorylated forms (50 kDa).
This gene encodes a transcription factor that was originally identified as a widely expressed mammalian DNA binding protein that could bind a tax-responsive enhancer element in the LTR of HTLV-1. The encoded protein was also isolated and characterized as the cAMP-response element binding protein 2 (CREB-2). The protein encoded by this gene belongs to a family of DNA-binding proteins that includes the AP-1 family of transcription factors, cAMP-response element binding proteins (CREBs) and CREB-like proteins. These transcription factors share a leucine zipper region that is involved in protein-protein interactions, located C-terminal to a stretch of basic amino acids that functions as a DNA binding domain. Two alternative transcripts encoding the same protein have been described. Two pseudogenes are located on the X chromsome at q28 in a region containing a large inverted duplication.Specifications
ATF4 | |
Monoclonal | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P18848, Q06507 | |
ATF4 | |
ATF4 Fusion Protein Ag1279 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3E4C5 | |
Unconjugated | |
ATF4 | |
ATF4, CREB 2, CREB2, DNA binding protein TAXREB67, TAXREB67, TXREB | |
Mouse | |
Protein G | |
RUO | |
11911, 468 | |
-20°C | |
Liquid |