missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF4 Mouse anti-Human, Mouse, Clone: 3E4C5, Proteintech
Mouse Monoclonal Antibody
£130.00 - £339.00
Specifications
Antigen | ATF4 |
---|---|
Clone | 3E4C5 |
Concentration | 1.2 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16876985
|
Proteintech
60035-1-IG-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16866985
|
Proteintech
60035-1-IG-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is a mouse monoclonal antibody raised against full length human ATF4 antigen. The antibody recognizes the 38 kDa ATF4 protein and its phosphorylated forms (50 kDa).
This gene encodes a transcription factor that was originally identified as a widely expressed mammalian DNA binding protein that could bind a tax-responsive enhancer element in the LTR of HTLV-1. The encoded protein was also isolated and characterized as the cAMP-response element binding protein 2 (CREB-2). The protein encoded by this gene belongs to a family of DNA-binding proteins that includes the AP-1 family of transcription factors, cAMP-response element binding proteins (CREBs) and CREB-like proteins. These transcription factors share a leucine zipper region that is involved in protein-protein interactions, located C-terminal to a stretch of basic amino acids that functions as a DNA binding domain. Two alternative transcripts encoding the same protein have been described. Two pseudogenes are located on the X chromsome at q28 in a region containing a large inverted duplication.Specifications
ATF4 | |
1.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ATF4, CREB 2, CREB2, DNA binding protein TAXREB67, TAXREB67, TXREB | |
ATF4 | |
IgG1 | |
Protein G | |
Antibody |
3E4C5 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P18848, Q06507 | |
11911, 468 | |
ATF4 Fusion Protein Ag1279 | |
Primary | |
-20°C | |
ATF4 |