missing translation for 'onlineSavingsMsg'
Learn More

3M™ Aura™ Particulate Respirators 9300+ Series

Particulate respirators have an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media. Available in FFP1, FFP2, and FFP3 protection levels with or without valves, Innovative chin tab for easy adjustments and an embossed, sculpted upper panel helps reduce eyewear fogging.

£12.31 - £2275.00

Specifications

Certifications/Compliance (Europe) EN 149:2001 + A1:2009
Certifications (EN) EN 149:2001 + A1:2009
Color White
Design Type Flat-fold
Material Polypropylene
View More Specs

Products 11
Product Code Brand Filter Type With Exhalation Valve Assigned Protection Factor (APF) Model Price Quantity & Availability  
Product Code Brand Filter Type With Exhalation Valve Assigned Protection Factor (APF) Model Price Quantity & Availability  
11717574
View Documents
3M™
7000088725
FFP1 No 4 9310 N/A N/A N/A
10583051
View Documents
3M™
7000088723
FFP2 No 10 9320
£47.50
Pack of 20
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
12672545
View Documents
3M™
7000088724
FFP1 Yes 4 9312 N/A N/A N/A
12602555
View Documents
3M™
7000088812
FFP3 No 20 9330 N/A N/A N/A
11777574
View Documents
3M™
7000062381
FFP1 Yes 4 PF9312 N/A N/A N/A
11787574
View Documents Promotion Details
3M™
7000062383
FFP2 No 10 PF9320
£17.30
Pack of 5
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
11797574
View Documents
3M™
7000062382
FFP2 Yes 10 PF9322 N/A
N/A
11707584
View Documents Promotion Details
3M™
7000062384
FFP3 Yes 20 PF9332
£38.24
Pack of 5
In Stock
to see stock.
Edge
Add to basket
 
16523771
View Documents Promotion Details
3M™
7000089312
FFP3 Yes 20 9332+ P3 valved
£2250.00
Pack of 330
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
16684912
View Documents Promotion Details
3M™
7000088730
FFP2 Yes 10 9322+ P2 valved
£1200.00
Pack of 330
Estimated Shipment: 02-06-2025
to see stock.
Add to basket
 
16674902
View Documents Promotion Details
3M™
7100231525
FFP3 No 20 9330+ P3
£2275.00
Pack of 440
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
Description

Description

Particulate respirators have an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media. Available in FFP1, FFP2, and FFP3 protection levels with or without valves, Innovative chin tab for easy adjustments and an embossed, sculpted upper panel helps reduce eyewear fogging.

  • Provide comfortable protection against dusts and mists in a wide variety of industrial applications and other hazardous environments.
  • Offers FFP1, FFP2 and FFP3 protection.
  • 3-panel design allows for greater facial movement.
  • Sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging and improves eyewear compatibility.
  • Low resistance filter technology promotes easier breathing and the innovative chin tab allows simple adjustments.
  • Choose without valve style or with valve styles that feature 3M™ Cool Flow™ to reduce heat build-up and help provide comfortable protection in hot and humid conditions.
  • Individually packed in hygienic wrapping to prevent contamination before use.
  • Braided, colour-coded headbands help you identify the protection level offered: yellow for FFP1 (APF 4), blue for FFP2 (APF 10) and red for FFP3 (APF 20).
  • Foldable design makes them easy to store when not in use.
  • Respirators work well with 3M eyewear and hearing protection products.
Specifications

Specifications

EN 149:2001 + A1:2009
White
Polypropylene
Polypropylene
Polyisoprene
Sculpted
Dusts and mists found in a wide variety of industrial applications and other work situations
3-panel
EN 149:2001 + A1:2009
Flat-fold
One Size
Yes
Aluminum, Polyurethene
Individually Packaged
Non-sterile
No
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 5 Years From Date of Manufacture

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Aura™ Particulate Respirators 9300+ Series

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.