missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Aura™ Particulate Respirator
3M™ Aura™ Particulate Respirator 9300+Gen3 Series. Making great things possible. Comfortably.
Brand: 3M™ 7100138357
210.00 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
- New 3M™ Cool Flow™ Comfort Valve allows a greater percentage of warm, exhaled air to escape, helping to increase your comfort. (compared to 3M™ Aura™ 9300+ equivalent model)
- 3M Advanced Electret (filter) Media helps you breathe easier
- Innovative tabs on the upper and lower panels simplify positioning and adjustment
- Braided headbands are color-coded (blue for FFP2, red for FFP3) for quick and easy identification
- Easy to store away with flat-fold design and individual wrapping
- FFP2 (NPF 12) breathing protection against hazardous work environments
- FFP3 (NPF 50) breathing protection against hazardous work environments
- Available in standard packs (20 per carton) or small packs (5 per carton)
- Made in the UK
Specifications
20 | |
CE Approved | |
Red | |
Single-use | |
Advanced Electret Media | |
One Size | |
FFP3 | |
9332+Gen3-SP |
EN 149 | |
EN 149:2001+A1:2009 FFP3 NR D | |
Flat-fold | |
FFP3 | |
General Purpose Particulate Respirator | |
Yes | |
Yes | |
Flat-fold |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction