missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Aura™ Particulate Respirator
3M™ Aura™ Particulate Respirator 9300+Gen3 Series. Making great things possible. Comfortably.
£170.00 - £347.78
Specifications
Certifications/Compliance | EN 149 |
---|---|
Design Type | Flat-fold |
Disposable | Single-use |
Material | Advanced Electret Media |
Product Type | General Purpose Particulate Respirator |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
17817603
|
3M™
7100138120 |
9322+Gen3-SP |
£170.00
Pack of 40 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
17897593
|
3M™
7100138357 |
9332+Gen3-SP |
£347.78
Pack of 40 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
Description
- Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
- New 3M™ Cool Flow™ Comfort Valve allows a greater percentage of warm, exhaled air to escape, helping to increase your comfort. (compared to 3M™ Aura™ 9300+ equivalent model)
- 3M Advanced Electret (filter) Media helps you breathe easier
- Innovative tabs on the upper and lower panels simplify positioning and adjustment
- Braided headbands are color-coded (blue for FFP2, red for FFP3) for quick and easy identification
- Easy to store away with flat-fold design and individual wrapping
- FFP2 (NPF 12) breathing protection against hazardous work environments
- FFP3 (NPF 50) breathing protection against hazardous work environments
- Available in standard packs (20 per carton) or small packs (5 per carton)
- Made in the UK
Specifications
EN 149 | |
Single-use | |
General Purpose Particulate Respirator | |
Yes | |
Flat-fold |
Flat-fold | |
Advanced Electret Media | |
One Size | |
Yes |