Learn More
Keystone Cleanroom Products™ Autoclavable Breather Bag for Isolators
These self-sealing breather bags offer a chevron peel and hang hole to suspend previously steam sterilized pouches in an isolator for VHP decontamination of outer packaging.
£620.00 - £3328.20
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | COC |
Closure Type | Self-seal |
Color | White |
Disposable | Single-use |
Product Code | Brand | Capacity (English) | Capacity (Metric) | Length (English) | Width (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Capacity (Metric) | Length (English) | Width (English) | Price | Quantity & Availability | ||
17766909
|
Keystone Cleanroom Products™
BBMGTVKSS0306HH |
15.1 oz. | 0.44 L | 6 in. | 3 in. |
£1165.00
Pack of 1000 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17756909
|
Keystone Cleanroom Products™
BBMGTVKSS02095HH |
16 oz. | 0.47 L | 9.5 in. | 2 in. |
£1340.00
Pack of 1000 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17786909
|
Keystone Cleanroom Products™
BBMGTVKSS03508HH |
23.5 oz. | 0.69 L | 8 in. | 3.5 in. |
£650.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17776909
|
Keystone Cleanroom Products™
BBMGTVKSS0308HH |
20.2 oz. | 0.6 L | 8 in. | 3 in. |
£620.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17716919
|
Keystone Cleanroom Products™
BBMGTVKSS0510HH |
41 oz. | 1.42 L | 10 in. | 5 in. |
£830.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17726919
|
Keystone Cleanroom Products™
BBMGTVKSS0608HH |
41 oz. | 1.42 L | 8 in. | 6 in. |
£810.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17796909
|
Keystone Cleanroom Products™
BBMGTVKSS0415HH |
50.4 oz. | 1.49 L | 15 in. | 4 in. |
£1125.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17706919
|
Keystone Cleanroom Products™
BBMGTVKSS0422HH |
66 oz. | 1.95 L | 22 in. | 4 in. |
£1125.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17736919
|
Keystone Cleanroom Products™
BBMGTVKSS0612HH |
67 oz. | 1.98 L | 12 in. | 6 in. |
£1125.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17786919
|
Keystone Cleanroom Products™
BBMGTVKSS0810HH |
67.2 oz. | 1.99 L | 10 in. | 8 in. |
£1180.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17776919
|
Keystone Cleanroom Products™
BBMGTVKSS0713HH |
76.4 oz. | 2.26 L | 13 in. | 7 in. |
£1180.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17796919
|
Keystone Cleanroom Products™
BBMGTVKSS0815HH |
100.8 oz. | 2.98 L | 15 in. | 8 in. |
£1345.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17726929
|
Keystone Cleanroom Products™
BBMGTVKSS1012HH |
100.8 oz. | 2.98 L | 11 in. | 10 in. |
£1590.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17706929
|
Keystone Cleanroom Products™
BBMGTVKSS0818HH |
121 oz. | 3.56 L | 18 in. | 8 in. |
£1720.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17746929
|
Keystone Cleanroom Products™
BBMGTVKSS1214HH |
141 oz. | 4.2 L | 14 in. | 12 in. |
£1770.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17746919
|
Keystone Cleanroom Products™
BBMGTVKSS0628HH |
141 oz. | 4.2 L | 28 in. | 6 in. |
£1770.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17756919
|
Keystone Cleanroom Products™
BBMGTVKSS0632HH |
161.3 oz. | 4.8 L | 32 in. | 6 in. |
£2600.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17736929
|
Keystone Cleanroom Products™
BBMGTVKSS1020HH |
168 oz. | 4.97 L | 20 in. | 10 in. |
£1870.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17756929
|
Keystone Cleanroom Products™
BBMGTVKSS1217HH |
169.3 oz. | 5 L | 17 in. | 12 in. |
£1910.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17766919
|
Keystone Cleanroom Products™
BBMGTVKSS0635HH |
176.4 oz. | 5.2 L | 35 in. | 6 in. |
£2715.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17716929
|
Keystone Cleanroom Products™
BBMGTVKSS0834HH |
228.5 oz. | 6.76 L | 34 in. | 8 in. |
£2780.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17786929
|
Keystone Cleanroom Products™
BBMGTVKSS1617HH |
228.5 oz. | 6.76 L | 17 in. | 16 in. |
£2600.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17776929
|
Keystone Cleanroom Products™
BBMGTVKSS1520HH |
252 oz. | 7.45 L | 20 in. | 15 in. |
£2635.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17766929
|
Keystone Cleanroom Products™
BBMGTVKSS1424HH |
278.9 oz. | 8.25 L | 24 in. | 14 in. |
£3030.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17726939
|
Keystone Cleanroom Products™
BBMGTVKSS1822HH |
332.6 oz. | 9.8 L | 22 in. | 18 in. |
£3180.00
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17796929
|
Keystone Cleanroom Products™
BBMGTVKSS1626HH |
349 oz. | 10.3 L | 26 in. | 16 in. |
£3328.20
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17706939
|
Keystone Cleanroom Products™
BBMGTVKSS1632HH |
430 oz. | 12.7 L | 32 in. | 16 in. |
£2045.00
Pack of 250 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17716939
|
Keystone Cleanroom Products™
BBMGTVKSS1635HH |
470.4 oz. | 13.9 L | 35 in. | 16 in. |
£2295.00
Pack of 250 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17736939
|
Keystone Cleanroom Products™
BBMGTVKSS2426HH |
524.2 oz. | 15.5 L | 26 in. | 24 in. |
£2165.00
Pack of 250 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17746939
|
Keystone Cleanroom Products™
BBMGTVKSS2430HH |
605 oz. | 17.9 L | 30 in. | 24 in. |
£2245.00
Pack of 250 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17756939
|
Keystone Cleanroom Products™
BBMGTVKSS2435HH |
706 oz. | 20.9 L | 35 in. | 24 in. |
£2405.00
Pack of 250 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
Description
- Tyvek™ and polyethylene film construction for high strength and exceptional microbial barrier does not absorb or adsorb VHP
- Designed for steam sterilization, but also works with gamma irradiation, EtO and VHP
- Fiber-free peelable seals
- High clarity for product visibility
- Recommended for sterilization of pharmaceutical equipment
- Manufactured and packaged in a certified ISO 6 cleanroom
- Works with both Impulse and Continuous Band Sealers
- Improves the efficiency of Parts Preparation Department
Specifications
Autoclavable | |
Self-seal | |
Single-use | |
Tyvek | |
203 μm | |
Steam, EtO, Gamma Irradiation, VHP |
COC | |
White | |
Isolator | |
Non-sterile | |
Yes | |
8 mil |