Learn More
Cole-Parmer™ Parker Hannifin™ PFA Threaded Adapter Compression Fitting
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings.
Brand: Cole-Parmer™ 31320-08
This item is not returnable.
View return policy
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and use the nut to compress down to form a secure seal
- Superior chemical resistance with durable PFA
- Available in male and female varieties
- Sterilization through autoclave or ethylene oxide (ETO)
Specifications
Compression Fitting | |
PFA | |
1 Each | |
1/8 in. NPT(M) |
Autoclavable | |
1/4 tubing O.D. x 1/8 in. NPT(M) Thread | |
Autoclave or Ethylene Oxide |