Learn More
Cole-Parmer™ Autoclave Deodorizer
Deoderize your autoclave with these capsules. Cole-Parmer™ Autoclave Deodorizers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them.
Brand: Cole-Parmer™ 44468-28
694.45 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
- Lemon Scented Capsules
- Absorb and remove autoclave odors rather than just covering them
- Simply place one capsule in your autoclave or incubator with each load
- The heat and moisture dissolves the capsule and releases a clean, fresh scent
Specifications
Autoclave Deodorizers, Lemon; 40/Bottle, 6 Bottles/Cs | |
40/Bottle, 6 Bottles/Cs. |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Product Content Correction
Product Title
Cole-Parmer™ Autoclave Deodorizer > Autoclave Deodorizer
Spot an opportunity for improvement?Share a Content Correction