Learn More
Bel-Art™ OdoClave™ Deodorant Autoclave Pads
Neutralize autoclave odors with a pleasant fragrance
Brand: Bel-Art™ H13198-0001
Description
Neutralize autoclave odors with a pleasant fragrance
- Heat-release deodorant works automatically at autoclave temperatures
- Open pouch and place in bag prior to autoclaving
- Three-year shelf life or longer if stored in a sealed container
- 32mm (1.75 in.) square
- Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads
Order Info
Choose 1.25 in. (32mm) square regular-strength or 2.25 in. (5.7cm) square super-strength pad.
Specifications
Autoclave Capsule | |
Liquid | |
Non-sterile | |
Red | |
Autoclave |
No | |
Cherry | |
Autoclavable | |
Pad |
Safety and Handling
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H411-Toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : dipentene,(R)-p-mentha-1,8-diene, terpineol, lemon oil, orange terpenes, amyl cinnamal, 2,6-dimethyl
Your input is important to us. Please complete this form to provide feedback related to the content on this product.