Learn More
Thermo Scientific™ Automation Tips for BioTek™ Pipetting Systems
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Brand: Thermo Scientific™ 984-261
Code : Z4
Additional Details : Weight : 9.97000kg
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: BioTek Precision and Precision XS.
- Available in non-sterile, sterile and with Thermo Scientific™ ART™ filter technology
- Lot-tested during production on actual workstation for which they are created
- Low percentage coefficient of variation (%CV)
Specifications
50 μL | |
Sterile | |
Rack | |
BioTek Pipetting System |
Non-Filtered | |
Clear | |
Low Retention | |
BioTek |