Learn More
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
GREENER_CHOICE
Increase pipetting performance with these automation tips. Each pipette tip undergoes a rigorous 15-point quality control program to ensure consistency and accuracy achieved by superior tip straightness and a low coefficient of variation.
Brand:  Thermo Scientific™ 967-021
721.99 GBP valid until 2025-05-09
Use promo code "24143" to get your promotional price.
 View more versions of this product
Product Code. 13306068
  • £913.00 / Pack of 3840
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Features:
  • Available in nonsterile, sterile and sterile filtered
  • Validated on actual workstation for which tip is created
  • Low percentage coefficient of variation
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 μL
Sterile
Rack
Eppendorf epMotion
Sterile and PCR Ready
Filtered
Clear
Automation
Eppendorf epMotion
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System > Filtered automation Tips for epMotion™ Volume: 1000μL; Sterility: Sterile; Color: Clear; Unit Size: 10 magazines of 384 tips/case (3840 tips in total)