Learn More
Molecular Probes™ Avidin, FITC conjugate
Avidin, FITC conjugate
Brand: Molecular Probes™ A821
Code : 50
Additional Details : Weight : 0.00525kg
Description
Our avidin, fluorescein conjugate has excitation and emission maxima ∼494/518. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.Specifications
FITC (Fluorescein) | |
Store in freezer (-5 to -30°C) and protect from light. | |
Avidin |
1 Each | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.