Learn More
Axygen™ 165 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
GREENER_CHOICEBIOPHARMA
For use with Agilent/Velocity11 automation systems
Brand:  Axygen™ VTF-165-R-S
 View more versions of this product
Product Code. 12776759
  • £579.12 / Pack of 4800
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tip format: 96 tip
  • Tip volume range: 20 μL to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: 96LT, 8LT
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Specifications
165 μL
Sterile
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Automation, Round
50 Racks/Cs.
Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT
Filtered
Clear
DNase, Rnase Free
Rack
SLAS Rack
w/out Bottom Label
Polyethylene