Learn More
Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Beveled, Reference
Designed to fit a wide variety of single and multi-channel pipettes
Brand:  Axygen™ TR-222-C-L-STK
253.26 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Code : 68
Additional Details : Weight : 3.97000kg
 View more versions of this product
Product Code. 12581298
  • £277.00 / Pack of 4800
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-205-WB-C-L)
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore
Specifications
Specifications
Graduated pipet tip
Non-Filtered
Clear
Refill
96 Tips/Rack, 5 Racks/Stack, 10 Stacks/Cs
Universal Fit, Universal Fit
Polypropylene
1 to 200 μL
Non-sterile
DNase, RNase Free
Beveled, Low Retention
Single-use
Yes
10, 50, 100μL