Learn More
Axygen™ Automation Tips for Agilent™/Velocity11 VPrep™ and Bravo: 384 Tips/Rack
GREENER_CHOICE
Comprehensive line of tips for Agilent/Velocity11 automation systems
Brand:  Axygen™ VT-384-10-L-R
 View more versions of this product
Product Code. 15262345
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Tip format: 96 tip
  • Tip volume range: 20 μL to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: 96LT, 8LT
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
Order Info
Packaging:10 Racks/Pack, 5 Packs/Case
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 μL
Non-sterile
Autoclavable
Maxymum Recovery™ technology for ultra-low retention
Automation, Round
50 Racks/Cs.
Polypropylene
Non-filtered
Clear
DNase, RNase, and Pyrogen free
Rack
SBS Rack
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST