missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Tips for Agilent™/Velocity11 VPrep™ and Bravo: 384 Tips/Rack
GREENER_CHOICE

Comprehensive line of tips for Agilent/Velocity11 automation systems

£4144.98 - £6275.00

Specifications

Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
For Use With (Application) Maxymum Recovery™ technology for ultra-low retention
Format Rack
View More Specs

Products 10
Product Code Brand For Use With (Equipment) Tip Style Sterility Volume (Metric) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Tip Style Sterility Volume (Metric) Price Quantity & Availability  
15272345
GREENER_CHOICE
View Documents
Axygen™
VT-384-70-L-R
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST Automation, Round Non-sterile 70 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15561453
GREENER_CHOICE
View Documents
Axygen™
VT-384-70-L-R-S
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST Automation, Round Sterile 70 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15581453
GREENER_CHOICE
View Documents
Axygen™
VTF-384-50-L-R-S
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST Automation, Round Sterile 50 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15531453
GREENER_CHOICE
View Documents
Axygen™
VT-384-31-L-R-S
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST Automation, Round Sterile 31 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15264646
GREENER_CHOICE
View Documents
Axygen™
VT-384-31-L-R
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST Automation, Round Non-sterile 31 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15571453
GREENER_CHOICE
View Documents
Axygen™
VTF-384-15-L-R-S
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST Automation, Round Sterile 15 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15521453
missing translation for 'viewDocuments'
Axygen™
VT-384-10-L-R-S
Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST Automation, Round Sterile 10 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15262345
GREENER_CHOICE
missing translation for 'viewDocuments'
Axygen™
VT-384-10-L-R
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST Automation, Round Non-sterile 10 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15541453
GREENER_CHOICE
missing translation for 'viewDocuments'
Axygen™
VT-384-5-L-R
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST Automation, Round Non-sterile 5 μL
£4530.00
Pack of 19200
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
 
15551453
GREENER_CHOICE
missing translation for 'viewDocuments'
Axygen™
VT-384-5-L-R-S
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST Automation, Round Sterile 5 μL
£4960.00
Pack of 19200
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
 
Description

Description

  • Tip format: 96 tip
  • Tip volume range: 20 μL to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: 96LT, 8LT
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene

Order Info

Packaging:10 Racks/Pack, 5 Packs/Case

Specifications

Specifications

Clear
DNase, RNase, and Pyrogen free
Rack
Autoclavable
Maxymum Recovery™ technology for ultra-low retention
Automation, Round
Videos
SDS
Documents

Documents

Certificates