Learn More
Axygen™ Eppendorf™ Compatible Pipet Tips
Compatible with Eppendorf and Biohit pipets
Brand:  Axygen TE-1004-B-L-R-S
95.21 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : A0
Additional Details : Weight : 2.19600kg
 View more versions of this product
Product Code. 12545667
  • £130.00 / Pack of 1152
In Stock
to see stock.
Add to basket
Description
Description
  • Available in sterile or nonsterile forms
  • e-Beam irradiated to prevent product degradation
  • Maxymum recovery tips allows complete sample retrieval and greater tip-to-tip precision
  • Graduation marks at 110μL and 500μL, allows visual confirmation of pipettor draw volume
  • Volume: 1000μL
  • Color: Blue
Specifications
Specifications
1000 μL
Blue
Low Retention
Eppendorf
110 and 500μL
Sterile
Rack
Racked
Yes