missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Eppendorf™ Compatible Pipet Tips
Compatible with Eppendorf and Biohit pipets
£71.41 - £348.00
Specifications
Volume (Metric) | 1000 μL |
---|---|
Color | Blue |
For Use With (Equipment) | Eppendorf |
Graduated | Yes |
Graduations | 110 and 500μL |
Product Code | Brand | Tip Style | Volume (Metric) | Packaging | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tip Style | Volume (Metric) | Packaging | Sterility | Price | Quantity & Availability | ||
12545667
|
Axygen
TE-1004-B-L-R-S |
Low Retention | 1000 μL | Racked | Sterile |
£130.00
Pack of 1152 |
In Stock
Log in to see stock. |
||
12736619
|
Axygen
TE-1004-B-L-R |
Low Retention | 1000 μL | Racked | Non-sterile |
£348.00
Pack of 3456 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12511318
|
Axygen
TE-1004-B |
Eppendorf | 1000 μL | Bulk | Non-sterile |
£176.00
Pack of 5000 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12756619
|
Axygen
TE-1004-B-R |
Eppendorf | 1000 μL | Racked | Non-sterile |
£332.00
Pack of 3456 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12766619
|
Axygen
TE-1004-B-R-S |
Eppendorf | 1000 μL | Racked | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12565667
|
Axygen
TE-1004-B-R-S |
Eppendorf | 1000 μL | Racked | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12535667
|
Axygen
TE-1004-B-L-R |
Low Retention | 1000 μL | Racked | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12555667
|
Axygen
TE-1004-B-R |
Eppendorf | 1000 μL | Racked | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Available in sterile or nonsterile forms
- e-Beam irradiated to prevent product degradation
- Maxymum recovery tips allows complete sample retrieval and greater tip-to-tip precision
- Graduation marks at 110μL and 500μL, allows visual confirmation of pipettor draw volume
- Volume: 1000μL
- Color: Blue
Order Info
- Bulk packed: 500 Tips/Pack. 10 Packs/Cs.
- Racked: 96 Tips/Rack, 12 Racks/Pack, 3 Packs/Cs.
Specifications
1000 μL | |
Eppendorf | |
110 and 500μL |
Blue | |
Yes |