missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Low Profile 8-Strip PCR Tubes
GREENER_CHOICE

Axygen™ brand low profile 8-strip PCR tubes are ideal for low sample volume reactions.

Brand:  Axygen™ PCR-0108-LP-RT-C

111.66 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15133707

  • £151.00 / Pack of 125

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Axygen™ Low Profile 8-Strip PCR tubes are ideal for low sample volume reactions. Using highly polished and precise molds, our tubes ensure precise thermal transfer and a leak-proof seal with a choice of strip caps. The low profile design allows reduced reaction time and evaporation due to less dead air volume. Tubes are available in white and clear with or without caps. Tubes are DNase-/RNase-free and nonpyrogenic.

 

  • Low profile design results in faster and more efficient PCR reactions
  • Ideal for 0.1 mL thermal cyclers
  • Available in white or clear
  • DNase-/RNase-free and Nonpyrogenic
  • Recommended volume range = 5 μL to 125 μL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RNase, DNase and non-pyrogenic free
0.1 mL
0.1 mL thermal cycler for low sample volume reactions
Non-sterile
Flat
Thin Wall 8-Strip PCR Tube with Real Time Strip Caps
Clear
125/Pk.
Conical
Polypropylene
Strip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers