missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ PCR Tubes with 0.2 mL Dome Cap
GREENER_CHOICE

Thin and consistent wall thickness for precise thermal transfer

Brand:  Axygen™ PCR-02D-A

 View more versions of this product

Product Code. 11311924

  • £65.10 / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Axygen™ 0.2 mL thin-wall 12 strip PCR tubes, available in an assortment of colors are designed to fit a wide variety of thermal cyclers. Ultra-thin and consistent wall thickness allows precise thermal transfer, even insertion cap ensures a tight fit to prevent evaporation.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Caps sold separately
  • Manufactured with 99.9% pure virgin polypropylene
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    RNase, DNase, pyrogen-free
    0.2 mL
    Polypropylene
    Conical
    Polypropylene
    Tube
    Assorted
    1000/Pk., 10Pk./Cs.
    Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
    Non-sterile
    Domed
    Thin Wall, Domed Cap PCR Tube
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers