missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Plate Sealer

Uniform and consistent sealing of microplates.

Brand:  Axygen HS-1230

Product Code. 12312099

  • £6495.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Adjustable sealing temperature: 100°C to 190°C
  • Compatible plate materials: PP (Polypropylene), PS (Polystyrene), PE (Polyethylene)
  • Compatible plate types: Standard assay plates, deep well storage plates, PCR plate
  • User-friendly control panel
  • Sealing operation is automated to ensure consistent results
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ideal for the low-to-medium throughput laboratory that requires uniform and consistent sealing of microplates
100° to 190°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ Plate Sealer > Voltage: 230V

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.