Learn More
Axygen™ Universal Fit Micropipette Tips
Axygen™ Universal Fit Pipette Tips are compatible with most research-grade single- and multi-channel micropipettes, and are certified to be DNase-/RNase-free and nonpyrogenic.
Brand:  Axygen™ TE-204-Y
327.74 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Code : 68
Additional Details : Weight : 2.72000kg
 View more versions of this product
Product Code. 12551348
  • £352.00 / Pack of 10000
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • RNase-/DNase free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Available in a variety of sizes
  • Available with a variety of tip styles including beveled, fine point, and wide bore
  • Available sterile/non-sterile
  • Available in bulk, racked, or stack rack packaging
  • Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery
Order Info
CapacityMetric
Specifications
Specifications
200 μL
Non-sterile
Autoclavable
Rack
Bulk
Universal Fit, Universal Fit
Polypropylene
Non-Filtered
Yellow
Dnase-, Rnase-, Pyrogen-, Human gDNA-, PCR-Inhibition-Free
Universal
500 tips/pack, 20 packs/case
Yes
10, 50, 100μL