missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Universal Fit Micropipette Tips
Axygen™ Universal Fit Pipette Tips are compatible with most research-grade single- and multi-channel micropipettes, and are certified to be DNase-/RNase-free and nonpyrogenic.
£37.93 - £443.00
Specifications
Volume (Metric) | 200 μL |
---|---|
Filtered | Non-Filtered |
Autoclavable | Autoclavable |
Clean Claims | Dnase-, Rnase-, Pyrogen-, Human gDNA-, PCR-Inhibition-Free |
Format | Rack |
Product Code | Brand | Color | Sterility | Tip Style | Packaging | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Sterility | Tip Style | Packaging | Price | Quantity & Availability | ||
12511308
|
Axygen™
TR-222-Y-STK |
Yellow | Non-sterile | Beveled | Stack Rack |
£238.00
Pack of 4800 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12655876
|
Axygen™
TR-222-C-STK |
Clear | Non-sterile | Beveled | Stack Rack |
£238.00
Pack of 4800 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12713119
|
Axygen™
T-205-WB-C-R |
Clear | Non-sterile | Genomic/Wide Orifice | Stack Rack |
£321.00
Pack of 4800 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12501298
|
Axygen™
T-200-C-STK-S |
Clear | Non-sterile | Beveled | Stack Rack |
£298.00
Pack of 4800 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12501308
|
Axygen™
TR-222-C-STK-S |
Clear | Sterile | Beveled | Stack Rack |
£267.00
Pack of 4800 |
In Stock
Log in to see stock. |
||
13197384
|
Axygen™
TE-204-Y-L-R |
Yellow | Non-sterile | Universal | Rack |
£347.00
Pack of 4800 |
In Stock
Log in to see stock. |
||
12786619
|
Axygen™
TE-204-Y-L-R-S |
Yellow | Sterile | Universal | Rack |
£443.00
Pack of 4800 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12796619
|
Axygen™
TE-204-Y-R |
Yellow | Non-sterile | Universal | Rack |
£293.00
Pack of 4800 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12726729
|
Axygen™
TE-204-Y-R-S |
Yellow | Sterile | Universal | Rack |
£344.00
Pack of 4800 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12551348
|
Axygen™
TE-204-Y |
Yellow | Non-sterile | Universal | Bulk |
£352.00
Pack of 10000 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
11314275
|
Axygen™
TE-204-Y-L-R |
Yellow | Non-sterile | Universal | Rack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11334275
|
Axygen™
TE-204-Y-R |
Yellow | Non-sterile | Universal | Rack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11324275
|
Axygen™
TE-204-Y-L-R-S |
Yellow | Sterile | Universal | Rack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11344275
|
Axygen™
TE-204-Y-R-S |
Yellow | Sterile | Universal | Rack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11314255
|
Axygen™
T-205-WB-C-R |
Clear | Non-sterile | Genomic/Wide Orifice | Stack Rack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- RNase-/DNase free
- Nonpyrogenic
- Human gDNA- and PCR-inhibition-free
- Available in a variety of sizes
- Available with a variety of tip styles including beveled, fine point, and wide bore
- Available sterile/non-sterile
- Available in bulk, racked, or stack rack packaging
- Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery
Specifications
200 μL | |
Autoclavable | |
Rack |
Non-Filtered | |
Dnase-, Rnase-, Pyrogen-, Human gDNA-, PCR-Inhibition-Free | |
Polypropylene |