Learn More
Axygen™ Single Well Medium Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
Brand:  Axygen™ RES-SW384-FX-SI
Code : 68
Additional Details : Weight : 2.70000kg
 View more versions of this product
Product Code. 12537877
  • £376.00 / Pack of 25
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
  • Durable and cost effective
  • Various formats for multiple applications
Specifications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic
170 mL
Sterile
Individually Wrapped; 25/Cs.
Single-use
Clear
Polypropylene
Reagent Reservoir
384