missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
GREENER_CHOICEBIOPHARMA

For sample collection, in-vitro growth chambers and long term storage.

Brand:  Axygen™ P-96-450V-C

131.24 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 12507927

  • £188.00 / Pack of 50
Estimated Shipment: 21-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.

 

  • 500 μL deep well plate
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANSI, SLAS 1-2004
Without Lid
Autoclavable
Polypropylene
Non-sterile
Sample collection, in-vitro growth chambers and long term storage
-112°F
Standard
8 x 12
Clear
96
Conical Bottom (V-well)
4000xG
-80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number30824015 Certificate TypeCertificate of Quality Date28/01/2025 Catalog NumberP-96-450V-C
Lot Number28224004 Certificate TypeCertificate of Quality Date24/12/2024 Catalog NumberP-96-450V-C
Lot Number20724002 Certificate TypeCertificate of Quality Date24/12/2024 Catalog NumberP-96-450V-C
Lot Number20124033 Certificate TypeCertificate of Quality Date03/12/2024 Catalog NumberP-96-450V-C
Lot Number20724001 Certificate TypeCertificate of Quality Date26/11/2024 Catalog NumberP-96-450V-C
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ Storage Microplates > 96-well; 450 μL; No; V-bottom

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.