Learn More
Fisherbrand™ 96-Well Polypropylene Microplates
197.33 GBP valid until 2025-04-30
Use promo code "24163" to get your promotional price.
Description
- Polypropylene
- Minimum binding
- Resistant to many reagents
- Withstand temperatures from -80° to +121°C (-112° to +250°F)
- Automation-friendly design optimized for HTS

Specifications
Standard | |
Natural | |
96 | |
Round |
8 x 12 | |
Polypropylene | |
Non-sterile | |
For storage feature generous 500ÎĽL well volumes |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Fisherbrand 96 Well Polypropylene Microplates
-
Fisherbrand Advantage aspiration system
Special Offers
-
Fisherbrand
Fisherbrand Sweet Deals on Top Products
Get up to 60% off select Fisherbrand productsPromo Code: 24163Expires: 2025-04-30
Days
4Hours
2Minutes
25Seconds
47 -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.