missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates

For sample collection, in-vitro growth chambers and long term storage.
Brand: Axygen™ P-5ML-48-C
204.76 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
Used for sample collection, in vitro growth chambers and long-term storage.
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
Specifications
ANSI, SLAS 1-2004 | |
6 x 8 | |
Clear | |
5000 μL | |
Non-sterile | |
Sample collection, in-vitro growth chambers and long term storage | |
25/Cs. | |
-112°F |
Standard | |
Autoclavable | |
Polypropylene | |
48 | |
Rectangular, Conical Bottom (V-well) | |
4000xG | |
5/Pk. | |
-80°C |
AXY001 Catalogue GENERAL AXYGEN FS
File size: 2.2 MB
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
7Hours
4Minutes
32Seconds
48
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ Storage Microplates > 48-well; 4.6 mL; No; Rectangular
Spot an opportunity for improvement?Share a Content Correction