missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Tip Refill System, Empty Racks

Easy-to-load refill racks for Axygen Tip Refill System. For use with P2/P10 Gilson-Style and 200μL Universal fit inserts.

£33.55 - £113.11

Specifications

Product Type Empty Refill Rack with Lid
Color Clear
Sterility Non-sterile
For Use With (Equipment) P2/P10 Gilson-Style and 200μL Universal fit inserts
View More Specs

Products 4
Product Code Brand Model Packaging Price Quantity & Availability  
Product Code Brand Model Packaging Price Quantity & Availability  
11330285
View Documents
Axygen™
RFL-R
RFL-R 10 Racks/Pack; 5 Packs/Case
£33.55
Pack of 10
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
 
12736719
View Documents
Axygen™
RFL-R
RFL-R 10 Racks/Pack; 5 Packs/Case This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11340285
View Documents Promotion Details
Axygen™
RFL-R-1200
RFL-R-1200 10 Racks/Pack; 3 Packs/Case
£43.90
Pack of 10
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
 
12746719
View Documents
Axygen™
RFL-R-1200
RFL-R-1200 10 Racks/Pack; 3 Packs/Case This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.

  • RFL-R is suitable for both the 10 μL and 200 μL refill inserts
  • RFL-R-1200 is suitable for 1200 μL refill inserts
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
Specifications

Specifications

Empty Refill Rack with Lid
Clear
Non-sterile
P2/P10 Gilson-Style and 200μL Universal fit inserts
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ Tip Refill System, Empty Racks

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.