Learn More
Axygen™ Tip Refill System, Empty Racks
Easy-to-load refill racks for Axygen Tip Refill System. For use with P2/P10 Gilson-Style and 200μL Universal fit inserts.
Brand:  Axygen™ RFL-R-1200
 View more versions of this product
Product Code. 12746719
Description
Description
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
  • RFL-R is suitable for both the 10 μL and 200 μL refill inserts
  • RFL-R-1200 is suitable for 1200 μL refill inserts
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Empty Refill Rack with Lid
Non-sterile
RFL-R-1200
Clear
P2/P10 Gilson-Style and 200μL Universal fit inserts
10 Racks/Pack; 3 Packs/Case
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ Tip Refill System, Empty Racks > 10 Racks/Pk, 5 Pk/Cs