missing translation for 'onlineSavingsMsg'
Learn More

Azadibenzocyclooctyne-NHS ester, with 4 carbon linker, Thermo Scientific Chemicals

£105.53 - £298.49

Specifications

Color White to Gray
Sensitivity Moisture sensitive
Solubility Information Soluble in dimethyl formamide,terahydrofuran,dimethyl sulfocide and dichloromethane.
Formula Weight 402.4
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne-NHS ester, with 4 carbon linker
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15420347
SDS View Documents
Thermo Scientific Alfa Aesar
J64128.MB
25 mg
£105.53
25mg
Please to purchase this item. Need a web account? Register with us today!
15430347
SDS View Documents
Thermo Scientific Alfa Aesar
J64128.MC
100 mg
£298.49
100mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification. It is used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification. It is used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.

Solubility
Soluble in dimethyl formamide, terahydrofuran, dimethyl sulfocide and dichloromethane.

Notes
Moisture and heat sensitive. Store in cool place. Incompatible with strong oxidizng agents.
Specifications

Specifications

White to Gray
Soluble in dimethyl formamide,terahydrofuran,dimethyl sulfocide and dichloromethane.
Solid
Moisture sensitive
402.4
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

missing translation for 'storageNote' : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.