Learn More
Tocris Bioscience™ AZD 8055
Highly potent and selective mTOR inhibitor
Brand:  Tocris Bioscience™ 7840/10
Additional Details : CAS Number : 1009298-09-2 Weight : 0.00970kg
 View more versions of this product
Product Code. 30000023
  • £101.51 / 10mg
Estimated Shipment: 05-08-2024
to see stock.
Description
Description
AZD 8055 is a highly potent, selective and ATP-competitive mTOR inhibitor (IC50 = 0.8 nM). Inhibits growth and induces autophagy in a range of cancer cell lines and xenografts, including NSCLC and glioblastoma (IC50 values are 20, 50 and 53 nM in H838, A549 and U87MG cell lines, respectively). Orally bioavailable.
Specifications
Specifications
5-[2,4-Bis[(3S)-3-methyl-4-morpholinyl]pyrido[2,3-d]pyrimidin-7-yl]-2-methoxybenzenemethanol
10 mg
C25H31N5O4
1009298-09-2
mTOR Inhibitors
0.98