Learn More
STEMCELL Technologies AZD8055
mTOR pathway inhibitor; Inhibits mTOR
Brand:  STEMCELL Technologies 73004
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 17110162
  • £129.60 / 10mg
Estimated Shipment: 31-07-2024
to see stock.
Description
Description
  • AZD8055 is a potent, selective ATP-competitive inhibitor of mammalian target of rapamycin (mTOR), with an IC50 value of 0.8 nM (Chresta et al.) It is very specific with ∼1,000-fold selectivity for mTOR over all PI3K isoforms and exhibits no activity against a panel of 260 kinases at concentrations up to 10 μM (Chresta et al.)
Specifications
Specifications
1009298-09-2
10 mg
≥98%
CCG-168
C25H31N5O4