missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies AZD8055
mTOR pathway inhibitor; Inhibits mTOR
£51.30 - £129.60
Specifications
CAS | 1009298-09-2 |
---|---|
Synonym | CCG-168 |
Molecular Formula | C25H31N5O4 |
Purity | ≥98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17100162
|
STEMCELL Technologies
73002 |
1 mg |
£51.30
1mg |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17110162
|
STEMCELL Technologies
73004 |
10 mg |
£129.60
10mg |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- AZD8055 is a potent, selective ATP-competitive inhibitor of mammalian target of rapamycin (mTOR), with an IC50 value of 0.8 nM (Chresta et al.) It is very specific with ∼1,000-fold selectivity for mTOR over all PI3K isoforms and exhibits no activity against a panel of 260 kinases at concentrations up to 10 μM (Chresta et al.)
Specifications
1009298-09-2 | |
CCG-168 | |
C25H31N5O4 | |
≥98% |