missing translation for 'onlineSavingsMsg'
Learn More
Azelaic acid, 96%, Thermo Scientific Chemicals Product Code.: 11349208

Azelaic acid, 96%, Thermo Scientific Chemicals

Product Code. 11349208
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11349208

Brand: Thermo Scientific Alfa Aesar 042587.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Azelaic acid acts as an anti-inflammatory compound. It is also exhibits anti acne activity. It up regulates expression of ant oxidative enzymes. Azelaic acid also inhibits proliferation of melanocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-99-9
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00004432
InChI Key BDJRBEYXGGNYIS-UHFFFAOYSA-N
Synonym azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110Show MoreShow Less
PubChem CID 2266
ChEBI CHEBI:48131
IUPAC Name nonanedioic acid
SMILES C(CCCC(=O)O)CCCC(=O)O

Specifications

Melting Point ∼102°C
Boiling Point 286°C (100mmHg)
Beilstein 1101094
Merck Index 14,905
Solubility Information Soluble in water (2.4mg/ml at 20°C),ethanol (5%),DMSO,methanol (70mg/ml),and alcohol.
Formula Weight 188.22
Percent Purity 96%
Density 1.225 g/mL
Quantity 500 g
Flash Point 215°C (419°F)
Physical Form Crystalline
Chemical Name or Material Azelaic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5001I24T Certificate TypeCertificate of Analysis Date21/06/2023 Catalog Number042587.22,042587.36,042587
Lot Number5008N19N Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042587.22,042587.36
Lot Number5009R31N Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042587.22,042587.36
Lot NumberB01T016 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number042587.22,042587.36
Lot NumberD07Q28A Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number042587.22,042587.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azelaic acid, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.