Learn More
Azelaic acid, 96%, Thermo Scientific Chemicals
Description
Azelaic acid acts as an anti-inflammatory compound. It is also exhibits anti acne activity. It up regulates expression of ant oxidative enzymes. Azelaic acid also inhibits proliferation of melanocytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | ∼102°C |
Boiling Point | 286°C (100mmHg) |
Beilstein | 1101094 |
Merck Index | 14,905 |
Solubility Information | Soluble in water (2.4mg/ml at 20°C),ethanol (5%),DMSO,methanol (70mg/ml),and alcohol. |
Formula Weight | 188.22 |
Percent Purity | 96% |
Density | 1.225 g/mL |
Quantity | 100 g |
Flash Point | 215°C (419°F) |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
5001I24T | Certificate of Analysis | 21/06/2023 | 042587.22,042587.36,042587 |
5008N19N | Certificate of Analysis | 12/04/2022 | 042587.22,042587.36 |
5009R31N | Certificate of Analysis | 12/04/2022 | 042587.22,042587.36 |
D07Q28A | Certificate of Analysis | 12/11/2021 | 042587.22,042587.36 |
B01T016 | Certificate of Analysis | 12/11/2021 | 042587.22,042587.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.