Learn More
Azidotri-n-butyltin(IV), 95%, Thermo Scientific Chemicals
£95.50 - £355.00
Chemical Identifiers
CAS | 17846-68-3 |
---|---|
Molecular Formula | C12H27N3Sn |
Molecular Weight (g/mol) | 332.079 |
MDL Number | MFCD00216557 |
InChI Key | JKVRTUCVPZTEQZ-UHFFFAOYSA-N |
Synonym | tributyltin azide, azidotributyltin iv, azidotributyltin, azidotributyl tin, tri-n-butylazidotin, azido tributyl stannane, azido tributyltin, tri-butyltin azide, tributyl tin azide, tributylstannylazide |
PubChem CID | 4984872 |
IUPAC Name | azido(tributyl)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)N=[N+]=[N-] |
Description
Azidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAzidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.
Solubility
Miscible with toluene, hexane, acetonitrile and tetrahydrofuran.
Notes
Use in a fume hood. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
17846-68-3 | |
332.079 | |
JKVRTUCVPZTEQZ-UHFFFAOYSA-N | |
4984872 | |
CCCC[Sn](CCCC)(CCCC)N=[N+]=[N-] |
C12H27N3Sn | |
MFCD00216557 | |
tributyltin azide, azidotributyltin iv, azidotributyltin, azidotributyl tin, tri-n-butylazidotin, azido tributyl stannane, azido tributyltin, tri-butyltin azide, tributyl tin azide, tributylstannylazide | |
azido(tributyl)stannane |
Specifications
17846-68-3 | |
113°C (235°F) | |
[CH3(CH2)3]3SnN3 | |
UN2788 | |
Miscible with toluene,hexane,acetonitrile and tetrahydrofuran. | |
CCCC[Sn](CCCC)(CCCC)N=[N+]=[N-] | |
332.079 | |
332.06 | |
Azidotri-n-butyltin(IV) |
1.212 | |
C12H27N3Sn | |
MFCD00216557 | |
tributyltin azide, azidotributyltin iv, azidotributyltin, azidotributyl tin, tri-n-butylazidotin, azido tributyl stannane, azido tributyltin, tri-butyltin azide, tributyl tin azide, tributylstannylazide | |
JKVRTUCVPZTEQZ-UHFFFAOYSA-N | |
azido(tributyl)stannane | |
4984872 | |
95% |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only