Learn More
Azlon™ Gas Delivery Tube
Polypropylene construction. Azlon™ Gas Delivery Tube are ideal substitutes for fragile glass alternatives.
Brand:  Azlon™ DT005
 View more versions of this product
Product Code. 12693012
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Pre-bent rigid polypropylene delivery tubes
  • Unbreakable in normal use
  • Suitable for many types of gas collection experiments in schools and universities
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Double Bend Gas Delivery Tube
4 mm
6 mm
Many types of gas collection experiments in schools and universities
Double bend
Polypropylene
475 mm
135°C