Learn More
Azlon™ Industrial HDPE Funnel
Constructed of Polyethylene and unbreakable in normal use. Azlon™ Industrial HDPE Funnels are suitable for food contact applications in accordance with EU and USA regulations. Feature straight sided upper rims to prevent spillage.
Brand: Azlon FWC114
23.70 GBP valid until 2024-11-29
Use promo code "21718" to get your promotional price.
Description
Constructed of Polyethylene and unbreakable in normal use. Azlon™ Industrial HDPE Funnels are suitable for food contact applications in accordance with EU and USA regulations. Feature straight sided upper rims to prevent spillage.
- Rigid industrial funnels in polyethylene
- Unbreakable in normal use
- Feature straight sided upper rims to prevent spillage
Specifications
245mm | |
24mm | |
239 mm |
4000 mL | |
HDPE |