Learn More
Azlon™ Industrial HDPE Funnel
Constructed of Polyethylene and unbreakable in normal use. Azlon™ Industrial HDPE Funnels are suitable for food contact applications in accordance with EU and USA regulations. Feature straight sided upper rims to prevent spillage.
Brand:  Azlon FWC118
 View more versions of this product
Product Code. 12090773
  • £60.05 / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Constructed of Polyethylene and unbreakable in normal use. Azlon™ Industrial HDPE Funnels are suitable for food contact applications in accordance with EU and USA regulations. Feature straight sided upper rims to prevent spillage.
  • Rigid industrial funnels in polyethylene
  • Unbreakable in normal use
  • Feature straight sided upper rims to prevent spillage
TRUSTED_SUSTAINABILITY
Specifications
Specifications
380mm
36mm
HDPE
351 mm