missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ LDPE Printed Slope Shoulder Wash Bottles

Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. Azlon™ LDPE Printed Slope Shoulder Wash Bottles are indelibly printed with a choice of seven popular laboratory solvents, appropriate hazard symbols and CAS numbers.

Brand:  Azlon™ WGF531P

23.16 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 10287674

  • £62.70 / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Description

Description

Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. AzlonTM LDPE Printed Slope Shoulder Wash Bottles are indelibly printed with a choice of seven popular laboratory solvents, appropriate hazard symbols and CAS numbers.

  • Labeled wash bottles for easy identification of contents
  • Slim design enables comfortable handling and enhanced ease of use
  • Fitted with color-coded polypropylene closures
  • Closures feature precision jet-tip aperture for fine control
TRUSTED_SUSTAINABILITY
Specifications

Specifications

250 mL
Translucent
For easy identification of contents
LDPE
Red
Not Autoclavable
Narrow
152 mm
Acetone
152 mm
Yes
8.45 oz.
54mm
Red Polypropylene closure
5/PK
Yes
No
Round
Wash Bottle
Polypropylene
Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azlon™ LDPE Printed Slope Shoulder Wash Bottles > Red lid and print; Acetone label; Capacity: 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.