Learn More
Azlon™ LDPE Wide Neck Wash Bottles
Use these translucent plastic bottles for a variety of liquid storage and sampling applications. Azlon™ LDPE Wide Neck Wash Bottles come with color-coded one-piece PP closures that maintain their shape in use.
£34.40 - £81.15
Specifications
Material LDPE
Closure Included Yes
Mouth Wide
Closure Material Polypropylene
View More Specs
Products 16
Product Code Brand Capacity (Metric) Closure Color Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Color Price Quantity & Availability  
12039847
View Documents
Azlon
WGW234P
250 mL Blue
£59.00
Pack of 5
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
10451055
View Documents Promotion Details
Azlon
WGW230P
250 mL White
£58.30
Pack of 5
In Stock
to see stock.
Edge
Add to basket
12019817
View Documents Promotion Details
Azlon
WGW231P
250 mL Red
£58.30
Pack of 5
In Stock
to see stock.
Edge
Add to basket
12064857
View Documents
Azlon
WGW233P
250 mL Green
£56.45
Pack of 5
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
12044857
View Documents
Azlon
WGW232P
250 mL Yellow
£57.55
Pack of 5
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
12481041
View Documents
Azlon
WGW229P
250 mL Assorted
£34.40
Each
In Stock
to see stock.
Edge
Add to basket
10759555
View Documents Promotion Details
Azlon
WGW235P
500 mL Blue
£64.50
Pack of 5
In Stock
to see stock.
Edge
Add to basket
10238074
View Documents
Azlon
WGW236P
500 mL White
£62.75
Pack of 5
In Stock
to see stock.
Edge
Add to basket
12029837
View Documents Promotion Details
Azlon
WGW237P
500 mL Red
£62.75
Pack of 5
In Stock
to see stock.
Edge
Add to basket
12044867
View Documents
Azlon
WGW238P
500 mL Yellow
£66.50
Pack of 5
In Stock
to see stock.
Edge
Add to basket
12094767
View Documents
Azlon
WGW239P
500 mL Green
£66.15
Pack of 5
In Stock
to see stock.
Edge
Add to basket
12034827
View Documents
Azlon
WGW443P
1000 mL White
£78.80
Pack of 5
In Stock
to see stock.
Edge
Add to basket
10442215
View Documents
Azlon
WGW447P
1000 mL Green
£81.15
Pack of 5
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
12044697
View Documents
Azlon
WGW444P
1000 mL Yellow
£80.55
Pack of 5
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
10186064
View Documents
Azlon
WGW446P
1000 mL Blue
£79.85
Pack of 5
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
10707655
View Documents
Azlon
WGW445P
1000 mL Red
£42.70
Pack of 5
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
Description
Description
Use these translucent plastic bottles for a variety of liquid storage and sampling applications. AzlonTM LDPE Wide Neck Wash Bottles come with color-coded one-piece PP closures that maintain their shape in use.
  • Square shoulder wash bottles featuring a wide neck for easy spill free filling
  • Fitted with color coded one-piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Specifications
Specifications
LDPE
Yes
Wide
Polypropylene